Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD68/SR-D1 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC20401R100UG
This item is not returnable.
View return policy
Description
CD68/SR-D1 Monoclonal specifically detects CD68/SR-D1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
CD68/SR-D1 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
AAB25811 | |
CD68 | |
Mouse myeloma cell line NS0-derived recombinant human CD68/SR-D1 Asn22-Ile320 Accession # AAB25811 | |
RUO | |
968 | |
Human | |
IgG2b |
Flow Cytometry | |
298807 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD_antigen: CD68, CD68, CD68 antigen, CD68 molecule, DKFZp686M18236, GP110, LAMP4, macrophage antigen CD68, macrosialin, SCARD1, scavenger receptor class D, member 1, SRD1, SR-D1 | |
Mouse | |
100 μg | |
Primary | |
Detects human CD68/SR-D1 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction