Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD69 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB23591N100UG
This item is not returnable.
View return policy
Description
CD69 Monoclonal specifically detects CD69 in Human samples. It is validated for Flow Cytometry.Specifications
CD69 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q07108 | |
CD69 | |
Mouse myeloma cell line NS0-derived recombinant human CD69 Gly64-Lys199 Accession # Q07108 | |
RUO | |
969 | |
Human | |
IgG2a |
Flow Cytometry | |
298614 | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, EA-1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leu23, Leukocyte surface antigen Leu-23, MLR-3, p60 | |
Mouse | |
100 μg | |
Primary | |
Detects human CD69 in Western blots. In Western blots, no cross-reactivity with mouse CD69 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction