Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD72 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB5405N100UG
This item is not returnable.
View return policy
Description
CD72 Monoclonal specifically detects CD72 in Human samples. It is validated for Flow Cytometry.Specifications
CD72 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CD72, CD72 antigenB-cell differentiation antigen CD72, CD72 molecule, CD72b, Ly-19.2, Ly-32.2, Lyb-2, LYB2lyb-2 | |
Chinese hamster ovary cell line CHO-derived recombinant human CD72, Arg117-Asp359, Accession # P21854 | |
100 μg | |
Primary | |
Detects human CD72 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2a |
Flow Cytometry | |
982405 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P21854 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
971 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction