Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD72 Antibody, R&D Systems™

Sheep Antibody
Supplier: R&D Systems AF5405100
This item is not returnable.
View return policy
Description
CD72 specifically detects CD72 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
CD72 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CD72, CD72 antigenB-cell differentiation antigen CD72, CD72 molecule, CD72b, Ly-19.2, Ly-32.2, Lyb-2, LYB2lyb-2 | |
Mouse myeloma cell line NS0-derived recombinant human CD72 Arg117-Asp359 Accession # P21854 | |
100 μg | |
971 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Flow Cytometry | |
Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells | |
P21854 | |
Sheep | |
Affinity Purified | |
Primary | |
Detects human CD72 in direct ELISAs. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction