Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD8 alpha Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB3803T100UG
This item is not returnable.
View return policy
Description
CD8 Monoclonal antibody specifically detects CD8 in Human samples. It is validated for Flow CytometrySpecifications
CD8 | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CD_antigen: CD8a, CD8, CD8 antigen, alpha polypeptide (p32), CD8A, CD8a molecule, LEU2, Leu2 T-lymphocyte antigen, MAL, OKT8 T-cell antigen, p32, T cell co-receptor, T8 T-cell antigen, T-cell antigen Leu2, T-cell surface glycoprotein CD8 alpha chain, T-lymphocyte differentiation antigen T8/Leu-2 | |
Mouse myeloma cell line NS0-derived recombinant human CD8 alpha , Ser22-Val198, Accession # NP_741696 | |
100 μg | |
Primary | |
Reconstitution default not known | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2a |
Flow Cytometry | |
1033123 | |
Flow Cytometry 0.25-1 ug/106 cells | |
NP_741696 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
925 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction