Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD8 alpha APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1509A025
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1509A025 25 Tests
FAB1509A100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB1509A025 Supplier R&D Systems Supplier No. FAB1509A025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

CD8 Monoclonal specifically detects CD8 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD8
Applications Flow Cytometry
Classification Monoclonal
Clone 37006
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. NP_741969
Gene Alias CD8, CD8A, CD8a molecule, LEU2, p32
Gene Symbols CD8A
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD8 alpha Ser22-Val198 Accession # NP_741969
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 925
Test Specificity Detects human CD8 alpha in direct ELISAs. In direct ELISAs, no cross-reactivity with mouse CD8 alpha is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.