Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD81 MAb (Clone 454729), R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $479.50
Specifications
Antigen | CD81 |
---|---|
Clone | 454729 |
Dilution | Immunohistochemistry 5-25 μg/mL, Immunocytochemistry 8-25 μg/mL |
Applications | Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB46151SP
![]() |
R&D Systems
MAB46151SP |
25 μg |
Each for $200.50
|
|
|||||
MAB46151100
![]() |
R&D Systems
MAB46151100 |
100 μg |
Each for $479.50
|
|
|||||
Description
CD81 Monoclonal specifically detects CD81 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.Specifications
CD81 | |
Immunohistochemistry 5-25 μg/mL, Immunocytochemistry 8-25 μg/mL | |
Monoclonal | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CD81, CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, TSPAN28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7 | |
HEK293 human embryonic kidney cell line transfected with human CD81, Accession # P60033 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CD81 in direct ELISAs. |
454729 | |
Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Human | |
P60033 | |
975 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title