Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD82/Kai-1 Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB4616U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB4616U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB4616U100 Supplier R&D Systems Supplier No. FAB4616U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD82/Kai-1 Monoclonal specifically detects CD82/Kai-1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD82/Kai-1
Applications Flow Cytometry
Classification Monoclonal
Clone 423524
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. AAH01821
Gene Alias CD82, CD82 antigenC33, CD82 molecule, IA4, IA4C33 antigen, Inducible membrane protein R2, KAI1, KAI1GR15, kangai 1 (suppression of tumorigenicity 6, prostate; CD82 antigen (R2 leukocyteantigen, antigen detected by monoclonal and IA4)), Metastasis suppressor Kangai-1, R2, SAR2, SAR2tetraspanin-27, ST6, ST6tspan-27, Suppressor of tumorigenicity 6 protein, Tetraspanin-27, Tspan-27, TSPAN27, TSPAN274F9
Gene Symbols CD82
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CD82/Kai-1 Met1-Tyr267 Accession # AAH01821
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3732
Test Specificity Detects human CD82/Kai-1. Stains human CD82/Kai-1 transfectants but not irrelevant transfectants.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.