Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD84/SLAMF5 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
$200.50 - $410.00
Specifications
Antigen | CD84/SLAMF5 |
---|---|
Clone | 273508 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
CD84/SLAMF5 Monoclonal specifically detects CD84/SLAMF5 in Human samples. It is validated for Western Blot.Specifications
CD84/SLAMF5 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9UIB8.1 | |
8832 | |
Mouse myeloma cell line NS0-derived recombinant human CD84/SLAMF5 Lys22-Arg220 Accession # Q9UIB8.1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CD84/SLAMF5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse CD84 is observed. |
273508 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD84, CD84 antigen, CD84 antigen (leukocyte antigen), CD84 molecule, Cell surface antigen MAX.3, DKFZp781E2378, hCD84, hly9-beta, leucocyte differentiation antigen CD84, leukocyte antigen CD84, Leukocyte differentiation antigen CD84, Ly-9B, mCD84, Signaling lymphocytic activation molecule 5, SLAM family member 5, SLAMF5, SLAMF5LY9B | |
CD84 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title