Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD84/SLAMF5 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF1855
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF1855 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF1855 Supplier R&D Systems Supplier No. BAF1855
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

CD84/SLAMF5 Polyclonal specifically detects CD84/SLAMF5 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen CD84/SLAMF5
Applications Western Blot, Flow Cytometry
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells
Gene Accession No. Q9UIB8.1
Gene Alias CD84, CD84 antigen, CD84 antigen (leukocyte antigen), CD84 molecule, Cell surface antigen MAX.3, DKFZp781E2378, hCD84, hly9-beta, leucocyte differentiation antigen CD84, leukocyte antigen CD84, Leukocyte differentiation antigen CD84, Ly-9B, mCD84, Signaling lymphocytic activation molecule 5, SLAM family member 5, SLAMF5, SLAMF5LY9B
Gene Symbols CD84
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD84/SLAMF5 Lys22-Arg220 Accession # Q9UIB8.1
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8832
Test Specificity Detects human CD84/SLAMF5 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.