Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD9 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$184.70 - $464.00
Specifications
Antigen | CD9 |
---|---|
Clone | 2665FF |
Dilution | Western Blot 2 μg/mL, Simple Western 50 μg/mL |
Applications | Western Blot, Simple Western |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11464SP
![]() |
R&D Systems
MAB11464SP |
25 μg |
Each for $184.70
|
|
|||||
MAB11464100
![]() |
R&D Systems
MAB11464100 |
100 μg |
Each for $464.00
|
|
|||||
Description
CD9 Monoclonal antibody specifically detects CD9 in Human samples. It is validated for Western Blot, Simple WesternSpecifications
CD9 | |
Western Blot 2 μg/mL, Simple Western 50 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
BA2, BA-2/p24 antigen, BTCC-1, CD9, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24) | |
Chinese Hamster Ovary cell line, CHO-derived human CD9, Ser112-Ile195, Accession # P21926 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
2665FF | |
Western Blot, Simple Western | |
Unconjugated | |
Rabbit | |
Human | |
P21926 | |
928 | |
IgG | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title