Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CD97 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2529
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2529 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB2529 Supplier R&D Systems Supplier No. MAB2529
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD97 Isoform 1 Monoclonal specifically detects CD97 Isoform 1 in Human samples. It is validated for Western Blot, Flow Cytometry, Neutralization, CyTOF-ready.

Specifications

Antigen CD97 Isoform 1
Applications Western Blot, Flow Cytometry, Neutralization, CyTOF
Classification Monoclonal
Clone 380903
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Neutralization 0.5-2.5 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_001775.2
Gene Alias CD97, TM7LN1
Gene Symbols CD97
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD97 Gln21-Gln398 Accession # NP_001775.2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 976
Test Specificity Detects human CD97 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse CD97 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.