Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD97 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB2529A
This item is not returnable.
View return policy
Description
CD97 Isoform 1 Monoclonal specifically detects CD97 Isoform 1 in Human samples. It is validated for Flow Cytometry.Specifications
CD97 Isoform 1 | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD97, TM7LN1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
976 | |
Human | |
Purified |
Flow Cytometry | |
380903 | |
Flow Cytometry 10 uL/10^6 cells | |
NP_001775.2 | |
CD97 | |
Mouse myeloma cell line NS0-derived recombinant human CD97 Gln21-Gln398 Accession # NP_001775.2 | |
100 Tests | |
Primary | |
Detects human CD97 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse CD97 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction