Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals Human CD97 ELISA Kit (Colorimetric)
SDP

Catalog No. NB232696 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB232696 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB232696 Supplier Novus Biologicals Supplier No. NBP320177

Specifications

Assay Range 23.44-1500 pg/mL
Assay Sensitivity 15.80 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Serum
For Use With (Application) ELISA
Gene ID (Entrez) 976
Gene Symbol ADGRE5
Quantity 1 Kit
Research Discipline GPCR, Immunology
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym CD97 antigenseven-span transmembrane protein, CD97 molecule, Leukocyte antigen CD97, seven transmembrane helix receptor, seven-transmembrane, heterodimeric receptor associated with inflammation, TM7LN1
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.