Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CD99 Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB3968U100UG
missing translation for 'nonReturnableMessage'
missing translation for 'returnPolicyLabel'
Description
CD99 Monoclonal specifically detects CD99 in Human samples. It is validated for Flow Cytometry.Specifications
CD99 | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
12E7, CD99, CD99 antigenY homolog, CD99 molecule, E2 antigen, HBA71, MIC2, MIC2 (monoclonal 12E7), MIC2Y, MSK5X, pilr-1, PILR-L, Protein MIC2, surface antigen MIC2, T-cell surface glycoprotein E2 | |
Mouse myeloma cell line, NS0-derived human CD99, Asp23-Asp122, Accession # P14209 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
1021527 | |
Flow Cytometry 0.25-1 ug/106 cells | |
P14209 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4267 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction