Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CDCP1 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB26662N100UG
This item is not returnable.
View return policy
Description
CDCP1 Monoclonal specifically detects CDCP1 in Human samples. It is validated for Flow Cytometry.Specifications
CDCP1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_073753 | |
CDCP1 | |
Mouse myeloma cell line NS0-derived recombinant human CDCP1 isoform 1 Ala33-Leu666 Accession # NP_073753 | |
RUO | |
64866 | |
Human | |
IgG1 |
Flow Cytometry | |
309121 | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD318, CD318 antigen, CUB domain containing protein 1, CUB domain-containing protein 1, Membrane glycoprotein gp140, SIMA135, SIMA135TRASK, Subtractive immunization M plus HEp3-associated 135 kDa protein, Transmembrane and associated with src kinases | |
Mouse | |
100 μg | |
Primary | |
Detects human CDCP1 in direct ELISAs. Recognizes intact protein (aa 33-666) of human CDCP1. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction