Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CDCP1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB26662
This item is not returnable.
View return policy
Description
CDCP1 Monoclonal specifically detects CDCP1 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, CyTOF-ready.Specifications
CDCP1 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, Immunoprecipitation 25 ug/mL, CyTOF-ready | |
NP_073753 | |
CDCP1 | |
Mouse myeloma cell line NS0-derived recombinant human CDCP1 isoform 1 Ala33-Leu666 Accession # NP_073753 | |
100 μg | |
Primary | |
Detects human CDCP1 in direct ELISAs. Recognizes intact protein (aa 33-666) of human CDCP1. | |
Human | |
Purified |
Flow Cytometry, Immunoprecipitation, CyTOF | |
309121 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD318, CD318 antigen, CUB domain containing protein 1, CUB domain-containing protein 1, Membrane glycoprotein gp140, SIMA135, SIMA135TRASK, Subtractive immunization M plus HEp3-associated 135 kDa protein, Transmembrane and associated with src kinases | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
64866 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction