Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CDCP1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | CDCP1 |
---|---|
Clone | 309112 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB26661SP
![]() |
R&D Systems
MAB26661SP |
25 μg |
Each for $200.50
|
|
|||||
MAB26661
![]() |
R&D Systems
MAB26661 |
100 μg |
Each for $502.50
|
|
|||||
Description
CDCP1 Monoclonal specifically detects CDCP1 in Human samples. It is validated for Western Blot.Specifications
CDCP1 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
NP_073753 | |
64866 | |
Mouse myeloma cell line NS0-derived recombinant human CDCP1 isoform 1 Ala33-Leu666 Accession # NP_073753 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CDCP1 in direct ELISAs and Western blots. Recognizes an epitope on human CDCP1 found within the amino acid segment 369-666. |
309112 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD318, CD318 antigen, CUB domain containing protein 1, CUB domain-containing protein 1, Membrane glycoprotein gp140, SIMA135, SIMA135TRASK, Subtractive immunization M plus HEp3-associated 135 kDa protein, Transmembrane and associated with src kinases | |
CDCP1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title