Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cdk6 Antibody, Novus Biologicals™
Mouse Monoclonal Antibody
$205.85 - $459.22
Specifications
Antigen | Cdk6 |
---|---|
Clone | 1092920 |
Dilution | Western Blot 0.5 μg/mL, Simple Western 10 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL |
Applications | Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11636100
|
R&D Systems
MAB11636100 |
100 μg |
Each for $459.22
|
|
|||||
MAB11636SP
|
R&D Systems
MAB11636SP |
25 μg |
Each for $205.85
|
|
|||||
Description
Cdk6 Monoclonal antibody specifically detects Cdk6 in Human samples. It is validated for Western Blot,Simple Western,Immunohistochemistry,ImmunocytochemistrySpecifications
Cdk6 | |
Western Blot 0.5 μg/mL, Simple Western 10 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
CDKN6, Cell division protein kinase 6, cyclin-dependent kinase 6, EC 2.7.11, EC 2.7.11.22, PLSTIREMGC59692, Serine/threonine-protein kinase PLSTIRE | |
E. coli -derived recombinant human CDK-6, Met1-Ala326, Accession # Q00534 | |
Primary | |
Reconstitute lyophilized material at 0.2 mg/ml in sterile PBS. For liquid material, refer to CoA for concentration. |
1092920 | |
Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
Q00534 | |
1021 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°CC as supplied. 1 month, 2 to 8°CC under sterile conditions after reconstitution. 6 months, -20 to -70°CC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title