Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CDK8 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6809
This item is not returnable.
View return policy
Description
CDK8 Polyclonal specifically detects CDK8 in Human samples. It is validated for Immunohistochemistry.Specifications
CDK8 | |
Polyclonal | |
Unconjugated | |
P49336 | |
CDK8 | |
E. coli-derived recombinant human CDK8 Asn380-Tyr464 Accession # P49336 | |
100 μg | |
Primary | |
Detects human CDK8 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) CDK2, rhCDK4, and CDK6 is observed. | |
Human | |
IgG |
Immunohistochemistry | |
LYOPH | |
Immunohistochemistry 5-15 ug/mL | |
CDK8 protein kinase, Cell division protein kinase 8, cyclin-dependent kinase 8, EC 2.7.11, EC 2.7.11.22, EC 2.7.11.23, K35, Mediator complex subunit CDK8, Mediator of RNA polymerase II transcription subunit CDK8, MGC126074, MGC126075, Protein kinase K35 | |
Sheep | |
Affinity Purified | |
RUO | |
1024 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction