Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CDX4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $468.00
Specifications
Antigen | CDX4 |
---|---|
Clone | 322007 |
Concentration | LYOPH |
Dilution | Immunocytochemistry 8-25 ug/mL |
Applications | Immunocytochemistry |
Description
CDX4 Monoclonal specifically detects CDX4 in Human samples. It is validated for Immunocytochemistry.Specifications
CDX4 | |
LYOPH | |
Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
O14627 | |
1046 | |
E. coli-derived recombinant human CDX4 Met1-Thr170 Accession # O14627 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CDX4 in direct ELISAs. |
322007 | |
Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
caudal type homeo box transcription factor 4, caudal type homeobox 4, caudal type homeobox transcription factor 4, Caudal-type homeobox protein 4, homeobox protein CDX-4 | |
CDX4 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title