Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CEACAM-1/CD66a Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 2 publications

$214.00 - $2020.00

Specifications

Antigen CEACAM1/CD66a
Clone 283324
Dilution Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Applications Western Blot, Immunohistochemistry, ELISA
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB22441SP
GREENER_CHOICE
View Documents
R&D Systems
MAB22441SP
25 μg
Each for $214.00
Only null left
Add to Cart
 
MAB22441100
GREENER_CHOICE
View Documents
R&D Systems
MAB22441100
100 μg
Each for $538.00
Only null left
Add to Cart
 
MAB22441500
GREENER_CHOICE
View Documents
R&D Systems
MAB22441500
500 μg
Each for $2,020.00
Only null left
Add to Cart
 
Description

Description

CEACAM1/CD66a Monoclonal specifically detects CEACAM1/CD66a in Human samples. It is validated for Western Blot, Immunohistochemistry, ELISA Capture (Matched Antibody Pair).
Specifications

Specifications

CEACAM1/CD66a
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Monoclonal
Mouse
Human
P13688
634
Mouse myeloma cell line NS0-derived recombinant human CEACAM-1 Gln35-Gly428 Accession # P13688
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human CEACAM-1 in ELISAs and Western blots. In ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CD31, rhICAM-1, -2, -3, recombinant mouse MAdCAM-1, or rhVCAM-1 was observed. In sandwich ELISAs, no cross-reactivity with rhCEACAM-3, rhCEACAM-5, or rhCEACAM-6 was observed.
283324
Western Blot, Immunohistochemistry, ELISA
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
antigen CD66, BGP1, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, Biliary Glycoprotein 1, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen, Cea-1, CEACAM1, Hv-1, Hv-2, MHVR
CEACAM1
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.