Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CEACAM-19 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $420.50
Specifications
Antigen | CEACAM19 |
---|---|
Clone | 1032215 |
Dilution | Flow Cytometry 0.25 ug/106 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9397SP
![]() |
R&D Systems
MAB9397SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9397100
![]() |
R&D Systems
MAB9397100 |
100 μg |
Each for $420.50
|
|
|||||
Description
CEACAM19 Monoclonal antibody specifically detects CEACAM19 in Human samples. It is validated for Flow CytometrySpecifications
CEACAM19 | |
Flow Cytometry 0.25 ug/106 cells | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
carcinoembryonic antigen-related cell adhesion molecule 19, CEACAM19, CEAL1 | |
Human embryonic kidney cell HEK293-derived human CEACAM-19, Ala33-Gly157, Accession # Q7692-1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1032215 | |
Flow Cytometry | |
Unconjugated | |
Mouse | |
Human | |
Q7692-1 | |
56971 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title