Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CEACAM-20 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB9416R100UG
This item is not returnable.
View return policy
Description
CEACAM20 Monoclonal antibody specifically detects CEACAM20 in Human samples. It is validated for Flow CytometrySpecifications
CEACAM20 | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CEACAM20, GPAD9366, UNQ9366 | |
Human embryonic kidney cell HEK293-derived human CEACAM-20, Gln31-Gly450, Accession # Q6UY09-1 | |
100 μg | |
Primary | |
Reconstitution default not known | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG1 |
Flow Cytometry | |
1032339 | |
Flow Cytometry 0.25-1 ug/106 cells | |
Q6UY09-1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
125931 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction