Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CEACAM-4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4354
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4354 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4354 Supplier R&D Systems Supplier No. MAB4354
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CEACAM4 Monoclonal specifically detects CEACAM4 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen CEACAM4
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 568410
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O75871
Gene Alias carcinoembryonic antigen CGM7, carcinoembryonic antigen gene family member 7, carcinoembryonic antigen-related cell adhesion molecule 4, CEACAM4, CGM7, CGM7_HUMAN
Gene Symbols CEACAM4
Host Species Mouse
Immunogen E. coli-derived recombinant human CEACAM-4 Phe36-Val140 Accession # O75871
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1089
Test Specificity Detects human CEACAM-4 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human CEACAM-1, -3, -5, -6, -7, or -8 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.