Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CEACAM-6/CD66c Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB3934R100UG
This item is not returnable.
View return policy
Description
CEACAM6/CD66c Monoclonal specifically detects CEACAM6/CD66c in Human samples. It is validated for Flow Cytometry.Specifications
CEACAM6/CD66c | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P40199 | |
CEACAM6 | |
Mouse myeloma cell line NS0-derived recombinant human CEACAM-6/CD66c Lys35-Gly320 Accession # P40199 | |
RUO | |
4680 | |
Human | |
IgG2a |
Flow Cytometry | |
439424 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
carcinoembryonic antigen-related cell adhesion molecule 6, carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific crossreacting antigen), CD66c, CD66c antigen, CEACAM6, CEAL, NCA, NCACEAL, Non-specific crossreacting antigen, Normal cross-reacting antigen | |
Mouse | |
100 μg | |
Primary | |
Detects human CEACAM-6/CD66c in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CEACAM-1, -3, or -5 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction