Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CEACAM-7 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB44781T100UG
This item is not returnable.
View return policy
Description
CEACAM7 Monoclonal specifically detects CEACAM7 in Human samples. It is validated for Flow Cytometry.Specifications
CEACAM7 | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide. | |
Carcinoembryonic antigen CGM 2, Carcinoembryonic antigen CGM2, Carcinoembryonic antigen gene family member 2, Carcinoembryonic antigen related cell adhesion molecule 7, CEA, CEACAM 7, CEACAM7, CGM 2, CGM2 | |
Chinese Hamster Ovary cell line, CHO-derived recombinant human CEACAM-7 Thr36-Asn233 Accession # Q14002 | |
100 μg | |
1087 | |
Human | |
Supernatant |
Flow Cytometry | |
962703 | |
Flow Cytometry 0.25-1μg/106 cells | |
Q14002 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human CEACAM-7 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction