Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CEACAM-7 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB44781100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB44781100 100 μg
MAB44781SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB44781100 Supplier R&D Systems Supplier No. MAB44781100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CEACAM7 Monoclonal specifically detects CEACAM7 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, CyTOF-ready.

Specifications

Antigen CEACAM7
Applications CyTOF, ELISA, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 962703
Conjugate Unconjugated
Dilution Flow Cytometry 0.25μg/106 cells, ELISA, Immunohistochemistry 5-25μg/mL, CyTOF-ready
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gene Accession No. Q14002
Gene Alias Carcinoembryonic antigen CGM 2, Carcinoembryonic antigen CGM2, Carcinoembryonic antigen gene family member 2, Carcinoembryonic antigen related cell adhesion molecule 7, CEA, CEACAM 7, CEACAM7, CGM 2, CGM2
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line, CHO-derived recombinant human CEACAM-7 Thr36-Asn233 Accession # Q14002
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1087
Test Specificity Detects human CEACAM-7 in direct ELISAs.
Reconstitution Reconstitute at 0.5mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.