Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CEACAM-8/CD66b Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB4246S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB4246S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB4246S100 Supplier R&D Systems Supplier No. FAB4246S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CEACAM8/CD66b Monoclonal specifically detects CEACAM8/CD66b in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CEACAM8/CD66b
Applications Flow Cytometry
Classification Monoclonal
Clone 913542
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P31997
Gene Alias Carcinoembryonic antigen CGM6, carcinoembryonic antigen gene family member 6, carcinoembryonic antigen-related cell adhesion molecule 8, CD66b, CD66b antigen, CD67, CD67 antigen, CEACAM8, CGM6, NCA-95, Non-specific cross-reacting antigen NCA-95
Host Species Mouse
Immunogen E.coli-derived recombinant human CEACAM-8/CD66b Gln35-His141 Accession # P31997
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1088
Test Specificity Detects human CEACAM-8/CD66b in direct ELISAs and Western blots.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.