Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CEACAM-8/CD66b Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4246SP
This item is not returnable.
View return policy
Description
CEACAM8/CD66b Polyclonal specifically detects CEACAM8/CD66b in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
CEACAM8/CD66b | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Carcinoembryonic antigen CGM6, carcinoembryonic antigen gene family member 6, carcinoembryonic antigen-related cell adhesion molecule 8, CD66b, CD66b antigen, CD67, CD67 antigen, CEACAM8, CGM6, NCA-95, Non-specific cross-reacting antigen NCA-95 | |
Sheep | |
Affinity Purified | |
RUO | |
1088 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Flow Cytometry, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
P31997 | |
CEACAM8 | |
E. coli-derived recombinant human CEACAM-8/CD66b Gln35-His141 Accession # P31997 | |
25 μg | |
Primary | |
Detects human CEACAM-8/CD66b in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) CEACAM-3, rhCEACAM-6, and rhCEACAM-7 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction