Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CEBP alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7094
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7094 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7094 Supplier R&D Systems Supplier No. AF7094
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

CEBP alpha Polyclonal specifically detects CEBP alpha in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen CEBP alpha
Applications Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 5-15 ug/mL
Gene Accession No. P49715
Gene Alias C/EBP alpha, C/EBP-alpha, CCAAT/enhancer binding protein (C/EBP), alpha, CCAAT/enhancer-binding protein alpha, CEBP, CEBPA
Gene Symbols CEBPA
Host Species Sheep
Immunogen E. coli-derived recombinant human CEBP alpha Met1-Ala124 Accession # P49715
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1050
Test Specificity Detects human CEBP alpha in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) CEBP beta , rhCEBP gamma , rhCEBP delta , rhCEBP zeta , and rhCEBP epsilon is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.