Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CEBP epsilon Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6726SP
This item is not returnable.
View return policy
Description
CEBP epsilon Monoclonal specifically detects CEBP epsilon in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
CEBP epsilon | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Q15744 | |
CEBPE | |
E. coli-derived recombinant human CEBP epsilon Thr151-Arg210 Accession # Q15744 | |
25 μg | |
Primary | |
Detects human CEBP epsilon in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CEBP alpha, beta, delta, gamma, or zeta is observed. | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
695345 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C/EBP-epsilon, CCAAT/enhancer binding protein (C/EBP), epsilon, CCAAT/enhancer-binding protein epsilon, CEBPE, CRP1C/EBP epsilon | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1053 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction