Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CEBP epsilon Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6726
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6726 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6726 Supplier R&D Systems Supplier No. MAB6726
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CEBP epsilon Monoclonal specifically detects CEBP epsilon in Human samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen CEBP epsilon
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 695345
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q15744
Gene Alias C/EBP-epsilon, CCAAT/enhancer binding protein (C/EBP), epsilon, CCAAT/enhancer-binding protein epsilon, CEBPE, CRP1C/EBP epsilon
Gene Symbols CEBPE
Host Species Mouse
Immunogen E. coli-derived recombinant human CEBP epsilon Thr151-Arg210 Accession # Q15744
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1053
Test Specificity Detects human CEBP epsilon in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CEBP alpha, beta, delta, gamma, or zeta is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.