Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CELSR3 PE-conjugated Antibody, R&D Systems™

Catalog No. FAB7278P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB7278P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB7278P Supplier R&D Systems Supplier No. FAB7278P
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Flamingo homolog 1/CELSR3 Monoclonal specifically detects Flamingo homolog 1/CELSR3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Flamingo homolog 1/CELSR3
Applications Flow Cytometry
Classification Monoclonal
Clone 763103
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. Q9NYQ7
Gene Alias Cadherin family member 11, cadherin, EGF LAG seven-pass G-type receptor 3 (flamingo homolog, Drosophila), CDHF11, CDHF11RESDA1, EGFL1, EGFL1cadherin EGF LAG seven-pass G-type receptor 3, EGF-like protein 1, EGF-like-domain, multiple 1, epidermal growth factor-like 1, Epidermal growth factor-like protein 1, Flamingo homolog 1, FMI1anchor protein, HFMI1, KIAA0812, MEGF2, MEGF2cadherin EGF LAG seven-pass G-type receptor 3, flamingo (Drosophila) homolog, multiple EGF-like domains 2, Multiple EGF-like domains protein 2, Multiple epidermal growth factor-like domains protein 2, RESDA1
Gene Symbols CELSR3
Host Species Mouse
Immunogen E. coli-derived recombinant human CELSR3 Val531-Ser711 Accession # Q9NYQ7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 1951
Test Specificity Detects human CELSR3 in ELISA.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.