Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CFTR R Domain Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 11 publications
Supplier: R&D Systems MAB1660SP
This item is not returnable.
View return policy
Description
CFTR Monoclonal specifically detects CFTR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunofluorescence.Specifications
CFTR | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 1-2 ug/10^6 cells, Immunofluorescence | |
P13569 | |
CFTR | |
beta-galactosidase-coupled CFTR Cys590-Lys830 (predicted) Accession # P13569 | |
25 μg | |
Primary | |
Detects human CFTR. Specifically recognizes in vitro synthesized CFTR, recombinant CFTR protein, and non-recombinant CFTR protein (1, 2). | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunofluorescence | |
13-1 | |
Unconjugated | |
Supplied as a solution in PBS containing BSA. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ABC35, ABCC7CF, ATP-binding cassette sub-family C member 7, ATP-binding cassette transporter sub-family C member 7, cAMP-dependent chloride channel, CFTR/MRP, Channel conductance-controlling ATPase, cystic fibrosis transmembrane conductance regulator, cystic fibrosis transmembrane conductance regulator (ATP-binding cassettesub-family C, member 7), cystic fibrosis transmembrane conductance regulator, ATP-binding cassette(sub-family C, member 7), dJ760C5.1, EC 3.6.3, MRP7EC 3.6.3.49, TNR-CFTR | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1080 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction