Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CGRP Antibody, Novus Biologicals™

Mouse Monoclonal Antibody
$177.10 - $464.50
Specifications
Antigen | CGRP1 |
---|---|
Clone | 1057808 |
Dilution | Western Blot 0.5 ug/mL, Immunohistochemistry 2-15 ug/mL, Immunocytochemistry 8-25 ug/mL |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11293100
![]() |
R&D Systems
MAB11293100 |
100 μg |
Each for $464.50
|
|
|||||
MAB11293SP
![]() |
R&D Systems
MAB11293SP |
25 μg |
Each for $177.10
|
|
|||||
Description
CGRP1 Monoclonal antibody specifically detects CGRP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunocytochemistrySpecifications
CGRP1 | |
Western Blot 0.5 ug/mL, Immunohistochemistry 2-15 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
a-CGRP, alpha-CGRP, alpha-type CGRP, CALCA, calcitonin gene-related peptide I, calcitonin/calcitonin-related polypeptide, alpha, calcitonin; calcitonin gene-related peptide 1, CGRP 1, CGRP alpha, CGRP I, CGRP-I | |
Human CGRP synthetic peptide (Tocris Catalog # 3012), Accession # P10092 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1057808 | |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P10092 | |
796 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title