Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CGRP2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB11323100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB11323100 100 μg
MAB11323SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB11323100 Supplier R&D Systems Supplier No. MAB11323100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CGRP2 Monoclonal antibody specifically detects CGRP2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry

Specifications

Antigen CGRP2
Applications ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 1057815
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Gene Accession No. P10092
Host Species Mouse
Immunogen Human CGRP synthetic peptide (Tocris Catalog # 3012), Accession # P10092
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 797
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.