Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CGRP2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$165.20 - $485.00
Specifications
Antigen | CGRP2 |
---|---|
Clone | 1057805 |
Dilution | Western Blot 1 ug/mL, ELISA, Immunocytochemistry 8-25 ug/mL |
Applications | Western Blot, ELISA, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11382100
![]() |
R&D Systems
MAB11382100 |
100 μg |
Each for $485.00
|
|
|||||
MAB11382SP
![]() |
R&D Systems
MAB11382SP |
25 μg |
Each for $165.20
|
|
|||||
Description
CGRP2 Monoclonal antibody specifically detects CGRP2 in Human samples. It is validated for Western Blot, ELISA, ImmunocytochemistrySpecifications
CGRP2 | |
Western Blot 1 ug/mL, ELISA, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
797 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
1057805 | |
Western Blot, ELISA, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P10092 | |
Human CGRP synthetic peptide (Tocris (Catalog # 3012 ), Accession # P10092 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title