Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CHD1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6195SP
This item is not returnable.
View return policy
Description
CHD1 Monoclonal specifically detects CHD1 in Human samples. It is validated for Western Blot, Immunocytochemistry, Knockout Validated.Specifications
CHD1 | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunocytochemistry 8-25 ug/mL, Knockout Validated | |
O14646 | |
CHD1 | |
E. coli-derived recombinant human CHD1 Lys1531-Thr1710 Accession # O14646 | |
25 μg | |
Primary | |
Detects human CHD1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human CHD1L, 5, or 7 is observed. | |
Human | |
Purified |
Western Blot, Immunocytochemistry, Immunoassay | |
677616 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ATP-dependent helicase CHD1, CHD-1, chromodomain helicase DNA binding protein 1, chromodomain-helicase-DNA-binding protein 1, DKFZp686E2337, EC 3.6.1, EC 3.6.4.12 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1105 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction