Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CHD1L Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6959
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6959 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6959 Supplier R&D Systems Supplier No. AF6959
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

CHD1L Polyclonal specifically detects CHD1L in Human samples. It is validated for Western Blot.

Specifications

Antigen CHD1L
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q86WJ1
Gene Alias ALC1, amplified in liver cancer 1, amplified in liver cancer protein 1, CHDL, chromodomain helicase DNA binding protein 1-like, chromodomain-helicase-DNA-binding protein 1-like, FLJ22530
Gene Symbols CHD1L
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human CHD1L Arg759-Lys879 Accession # Q86WJ1
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9557
Test Specificity Detects human CHD1L in direct ELISAs and Western blots. In direct ELISAs, less than 3% cross-reactivity with recombinant human (rh) CHD1 and rhCHD5 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.