Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CHD7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7350
This item is not returnable.
View return policy
Description
CHD7 Monoclonal specifically detects CHD7 in Human samples. It is validated for Immunocytochemistry.Specifications
CHD7 | |
Monoclonal | |
LYOPH | |
Immunocytochemistry 8-25 ug/mL | |
Q9P2D1 | |
CHD7 | |
E. coli-derived recombinant human CHD7 Ala263-Gln457 Accession # Q9P2D1 | |
100 μg | |
Primary | |
Detects human CHD7 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human CEBP epsilon is observed. | |
Human | |
Purified |
Immunocytochemistry | |
772503 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ATP-dependent helicase CHD7, CHD-7, chromodomain helicase DNA binding protein 7, chromodomain helicase DNA binding protein 7 isoform CRA_e, chromodomain-helicase-DNA-binding protein 7, EC 3.6.1, EC 3.6.4.12, FLJ20357, FLJ20361, IS3, KIAA1416KAL5 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
55636 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction