Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CHD7 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7350
This item is not returnable.
View return policy
Description
CHD7 Polyclonal specifically detects CHD7 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
CHD7 | |
Polyclonal | |
Unconjugated | |
Q9P2D1 | |
CHD7 | |
E. coli-derived recombinant human CHD7 Ala263-Gln457 Accession # Q9P2D1 | |
100 μg | |
Primary | |
Detects human CHD7 in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunocytochemistry 5-15 ug/mL | |
ATP-dependent helicase CHD7, CHD-7, chromodomain helicase DNA binding protein 7, chromodomain helicase DNA binding protein 7 isoform CRA_e, chromodomain-helicase-DNA-binding protein 7, EC 3.6.1, EC 3.6.4.12, FLJ20357, FLJ20361, IS3, KIAA1416KAL5 | |
Sheep | |
Affinity Purified | |
RUO | |
55636 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction