Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Chemerin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB23241SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB23241SP 25 μg
MAB23241100 100 μg
MAB23241500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB23241SP Supplier R&D Systems Supplier No. MAB23241SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 3 publications

Chemerin Monoclonal specifically detects Chemerin in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Chemerin
Applications ELISA
Classification Monoclonal
Clone 365314
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q99969
Gene Alias chemerin, RARRES2, RAR-responsive protein TIG2, retinoic acid receptor responder (tazarotene induced) 2, retinoic acid receptor responder protein 2, Tazarotene-induced gene 2 protein, TIG-2, TIG2HP10433
Gene Symbols RARRES2
Host Species Mouse
Immunogen E. coli-derived recombinant human Chemerin Glu21-Ser157 Accession # Q99969
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5919
Test Specificity Detects human Chemerin in ELISAs. In ELISAs, this antibody does not cross-react with recombinant mouse Chemerin.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.