Learn More
Human ChemR23 Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Supplier:  R&D Systems FAB362S100UG
Catalog No. FAB362S100
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
ChemR23/CMKLR1 Monoclonal specifically detects ChemR23/CMKLR1 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
ChemR23/CMKLR1
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
NP_004063
CMKLR1
NS0 mouse myeloma cell line transfected with human ChemR23 Met1-Leu371 Accession # NP_004063
RUO
1240
Human
IgG3
Flow Cytometry
84939
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CHEMERINR, chemokine receptor-like 1, chemokine-like receptor 1, CHEMR23, cmklr1, Dez, Gpcr27, G-protein coupled receptor ChemR23, G-protein coupled receptor DEZ, MGC126105, MGC126106, orphan G-protein coupled receptor, Dez
Mouse
100 μg
Primary
Detects human ChemR23. Stains human ChemR23-transfected cells but not irrelevant transfectants.
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only (RUO)