Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Chitinase 3-like 2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5116
This item is not returnable.
View return policy
Description
Chitinase 3-like 2 Monoclonal specifically detects Chitinase 3-like 2 in Human samples. It is validated for Western Blot.Specifications
| Chitinase 3-like 2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q15782 | |
| CHI3L2 | |
| Mouse myeloma cell line NS0-derived recombinant human Chitinase 3-like 2 Tyr27-Leu390 Accession # Q15782 | |
| 100 μg | |
| Primary | |
| Detects human Chitinase 3-like 2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) Chitinase 3-like 1 or rhChitotriosidase is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 632010 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CHI3L2, Chitinase 3 like 2, chitinase 3-like 2, chitinase-3-like protein 2, Chondrocyte protein 39, YKL39, YKL-39YKL39 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1117 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction