Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Chk2 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $467.50
Specifications
Antigen | Chk2 |
---|---|
Clone | 2521A |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1358SP
![]() |
R&D Systems
MAB1358SP |
25 μg |
Each for $200.50
|
|
|||||
MAB1358100
![]() |
R&D Systems
MAB1358100 |
100 μg |
Each for $467.50
|
|
|||||
Description
Chk2 Monoclonal specifically detects Chk2 in Human samples. It is validated for Western Blot.Specifications
Chk2 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CDS1, CHEK2, CHK2 checkpoint homolog (S. pombe), EC 2.7.11, EC 2.7.11.1, HuCds1, LFS2, PP1425, Rad53, S.pombe) homolog | |
E. coli-derived human Chk2, Asp208-Leu543, Accession # O96017 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
2521A | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O96017 | |
11200 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title