Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human CHL1 Partial ORF (NP_006605, 26 a.a. - 135 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89939489
Click to view available options
Quantity:
10 μg
25 μg

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the L1 gene family of neural cell adhesion molecules. It is a neural recognition molecule that may be involved in signal transduction pathways. The deletion of one copy of this gene may be responsible for mental defects in patients with 3p- syndrome. Several alternatively spliced transcript variants of this gene have been described, but their full length nature is not known. [provided by RefSeq]

Sequence: EIPSSVQQVPTIIKQSKVQVAFPFDEYFQIECEAKGNPEPTFSWTKDGNPFYFTDHRIIPSNNSGTFRIPNEGHISHFQGKYRCFASNKLGIAMSEEIEFIVPSVPKFPK

Specifications

Accession Number NP_006605
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10752
Molecular Weight (g/mol) 37.84kDa
Name CHL1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen EIPSSVQQVPTIIKQSKVQVAFPFDEYFQIECEAKGNPEPTFSWTKDGNPFYFTDHRIIPSNNSGTFRIPNEGHISHFQGKYRCFASNKLGIAMSEEIEFIVPSVPKFPK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CALL/FLJ44930/L1CAM2/MGC132578
Common Name CHL1
Gene Symbol CHL1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.