Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CHMP2B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7509
This item is not returnable.
View return policy
Description
CHMP2B Monoclonal specifically detects CHMP2B in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CHMP2B | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Q9UQN3 | |
CHMP2B | |
E. coli-derived recombinant human CHMP2B Ala2-Asp213 Accession # Q9UQN3 | |
100 μg | |
Primary | |
Detects human CHMP2B in direct ELISAs and Western blots | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
791521 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
charged multivesicular body protein 2b, CHMP2.5, CHMP2.5VPS2BVPS2-2, CHMP2b, chromatin modifying protein 2B, Chromatin-modifying protein 2b, DKFZP564O123, DMT1, hVps2-2, Vacuolar protein sorting-associated protein 2-2, vacuolar protein-sorting-associated protein 2-2, VPS2 homolog B, Vps2-2, VPS2B | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
25978 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction