Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Chondroadherin Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF8218
This item is not returnable.
View return policy
Description
Chondroadherin Polyclonal specifically detects Chondroadherin in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Chondroadherin | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CHAD, chondroadherin, chondroadherin proteoglycan, SLRR4A, SLRR4ACartilage leucine-rich protein | |
Sheep | |
Immunogen affinity purified | |
Primary | |
Detects human Chondroadherin in direct ELISAs and Western Blots. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 2 ug/mL, Immunohistochemistry 5-15 ug/mL | |
O15335 | |
CHAD | |
Mouse myeloma cell line NS0-derived recombinant Chondroadherin Cys23-His359 Accession # O15335 | |
100 μg | |
1101 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction