Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Chordin-like 1/CHRDL1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1808SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1808SP 25 μg
AF1808 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF1808SP Supplier R&D Systems Supplier No. AF1808SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Chordin-like 1/CHRDL1 Polyclonal specifically detects Chordin-like 1/CHRDL1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Chordin-like 1/CHRDL1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_001137454
Gene Alias CHL, chordin-like 1, chordin-like protein 1, CHRDL1, CRDL1, dA141H5.1, Neuralin 1, neuralin-1, Neurogenesin-1, NRLN1, NRLN1neuralin 1, ventroptin, Ventroptin alpha, VOPT
Gene Symbols CHRDL1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Chordin-like 1/CHRDL1 Glu28-Cys457 Accession # NP_001137454
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91851
Test Specificity Detects human Chordin-like 1/CHRDL1 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.